Stirring Shaft, Cresent Type, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Precision ground glass stirring shafts, 10mm O.D., crescent type
Useful for stirring viscous materials at low rpm. Available for round bottom flasks of 1, 3 and 5 liters. Catelog #89061-956, three liter size, will also fit a two liter flask, but closely. Flask opening must be a [ST] 29/42 joint or larger. Use with Trubore® bearings. Compatible with Ace PTFE mechanical seal and PTFE collet-type bearings. Join to stirrer motor using a chuck. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought