Cork Rings, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Fine-composition cork rings ideally suited for general laboratory use to support flasks, beakers, dishes
Rings are 30mm (1-3/16-inch) thick. Available in a variety of sizes.
Customers Who Bought This Also Bought