Lead-Free, Non-Aerated Eyesaver Faucet, Model SEF-1800NA, Speakman®
# de catalogue 97008-324
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Lead-Free, Non-Aerated Eyesaver Faucet, Model SEF-1800NA
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. no.:97008-324
- Finish:Polished Chrome and Yellow
- Supplier No.:SEF-1800-CA-NA
Spécifications
A propos de cet article
Speakman® Eyesaver® combination eyewashes and lavatory faucets are a revolutionary and exclusive product with a unique design.
- The faucet always remains functional
It combines a fully functional gooseneck faucet with an independently operated eyewash. This patented design incorporates two separate water channels in the faucet body, one for the eyewash and the other for the faucet. A simple pull activates the eyewash and delivers a safe water temperature to the eyes each and every time. The SEF-1800NA Model is a two-handle, widespread, chrome-plated, lead-free brass gooseneck faucet with integral non-aerated eyewash spray outlets.
Certifications: Meets ANSI Z358.1, NSF 61, CA AB1953, and VT Act 193.
Customers Who Bought This Also Bought