Eyesaver Drench Hose and Eyewash Combination, Model SE-920, Speakman®
# de catalogue VF-SE-920
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Eyesaver Drench Hose and Eyewash Combination, Model SE-920
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. no.:VF-SE-920
- Finish:Polished Chrome and Yellow
- Supplier No.:SE-920
Spécifications
A propos de cet article
Speakman® Drench hose and eyewash combinations provide great flexibility in laboratories and other industrial areas for flushing the eyes and body.
- Hose and eyewash combination
- Laboratory flexibility
- For direct flushing on contaminated areas
All units come standard with a flexible hose. Model SE-920 is a wall mounted drench hose featuring one aerated spray outlet and a 6' hose. The unit also features a self-closing valve and hook for the drench hose.
Certifications: Meets ANSI 358.1.
Customers Who Bought This Also Bought