Safe-T-Zone® Select Series Combination Shower, Model SE-616, Speakman®
# de catalogue VF-SE-616
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Safe-T-Zone® Select Series Combination Shower, Model SE-616
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. no.:VF-SE-616
- Inlet Size:1.25" NPT
- Supplier No.:SE-616
Spécifications
A propos de cet article
Speakman® Safe-T-Zone® Select Series combination shower and eyewash unit features a combination eyewash with two yellow aerated nylon spray outlets and a rectangular bowl.
- Showerhead is activated by a high visibility yellow pull rod
The outlets and bowl are combined on a DuraJade™ powder-coated, galvanized steel stanchion with an impeller action deluge showerhead. The Model SE-616 combination shower is supplied with an SE-505 eyewash with stainless steel bowl.
Certifications: Meets ANSI 358.1.
Customers Who Bought This Also Bought