Upchurch Scientific® Pressure Relief Valves, IDEX Health & Science
Fournisseur: Upchurch Scientific
Certificats
A propos de cet article
Prevent system over-pressurization with Upchurch Scientific® Pressure Relief Valves
To open the Pressure Relief Valve Assembly at a different pressure than 100psi (7 bar), simply combine one of the Upchurch Scientific® Replacement Back Pressure Regulator Assemblies with item 21511-160 Pressure Relief Valve Tee, or item 21528-160 for larger bore tubing and higher flow applications.
These products protect system components by diverting fluid flow automatically when inline pressure exceeds the set limit.
Certifications: ROHS certified.
Ordering information: The 21511-532 assembly and 21511-160 tee each ship with two 21511-110 nuts and two 21511-108 ferrules. The 21511-532 assembly also ships with wrenches to open/tighten the cartidge holder. To replace the cartidge supplied with 21511-532, order item 21511-588. The 21528-160 tee ships with two 21511-102 fittings and two 53498-548 ferrules.
Customers Who Bought This Also Bought