Laboratory Eyewash, Speakman®
# de catalogue 97034-678
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Laboratory Eyewash
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. no.:97034-678
- Finish:Polished Chrome
- Supplier No.:SE-573
Spécifications
A propos de cet article
Speakman® Laboratory products are designed to accommodate the needs of laboratories by offering a number of configurations to ensure the perfect fit for virtually any application.
- Includes a universal emergency sign
- Eyewash unit features two yellow spray heads
The Laboratory Eyewash unit minimizes the space needed for emergency equipment while offering maximum safety protection. The unit is mounted to the back of a deck surface and activated by being pulled downward.
Certifications: Meets ANSI 358.1
Customers Who Bought This Also Bought