Drench Hose Attachment, Speakman®
# de catalogue VF-4920-SE
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Color:Yellow and White
- Description:Drench Hose Attachment
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. no.:VF-4920-SE
- Supplier No.:SE-4920
Spécifications
A propos de cet article
The drench hose attachment is designed for use with the Speakman® portable GravityFlo® units.
This attachment allows for spot body washing, in addition to the unit's eyewash flushing capability.
Customers Who Bought This Also Bought