Commander® Service Sink Faucet with Cross Handles, Speakman®
# de catalogue 87003-002
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Faucet with Cross Handles
- Cat. no.:87003-002
- Supplier No.:SC-5811
Spécifications
A propos de cet article
The Speakman® Commander® service sink faucet line provides facilities with high quality and reliable brass faucets.
- The SC-5811 was designed for through the wall installations on 8" centers
- The faucet's threaded brass wall flanges provide for a +/- adjustments up to 1/8"
- Wall flanges and mounting screws are provided with the SC-5811
Ideal for janitorial closets, utility sinks, and other maintenance applications, the SC-5811 gooseneck faucet features a cast brass body and nozzle with 3/4" hose thread and pail hook and polished chrome plated brass finish. The service sink faucets feature 1/4 turn ceramic cartridges and a brass top brace assembly.
Certifications: The SC-5811 meets ASME A112.18.1/CSA B125.1 and ASSE-1001.
Customers Who Bought This Also Bought