Mighty Scope Dual View Stand, Aven Inc.
# de catalogue 89239-400
Fournisseur: Aven Tools
Spécifications
- Dimensions:30L x 20W x 30H cm (113/4 x 77/8 x 113/4 ")
- Rod Length:17 cm (6⅝") adjustable, 4.1 cm (1⅝") fine adjustment travel (arm)
- Description:Mighty Scope Dual View Stand, Working Distance: 29 cm (111/2")
- Cat. no.:89239-400
- Supplier No.:26700-215
Spécifications
A propos de cet article
The Dual View Stand allows users to mount two Mighty Scopes
Fits Mighty Scope 1.3M and 5M.
Plug these Mighty Scopes into a PC and use the MicroViewer Software (supplied with the Mighty Scope) to view two images at the same time. Excellent for viewing a specimen from different angles. The stand offers articulating arms that allows both up and down and back and forth movements, as well as rack and pinon focusing for fine adjustments.
Working distance: 29 cm (111/2")
Customers Who Bought This Also Bought