ContactUsLinkComponent
Mighty Scope Cradle Stand, Aven Inc.
Catalogue # 89239-396
Fournisseur:  Aven Tools
undefined
Mighty Scope Cradle Stand, Aven Inc.
Catalogue # 89239-396
Fournisseur:  Aven Tools
Supplier Number:  26700-213
Spécifications
  • Description:
    Mighty Scope Cradle Stand
  • Cat. no.:
    89239-396
  • Supplier No.:
    26700-213
Spécifications
À propos de cet article
Cradle stand includes base unit with two insert adaptors providing three “hands free” fixed working distance and magnification options. Ideal for collectors, hobbyists, and graphic artists, as well as in the home and classroom.
The stand is designed so that the Mighty Scope simply slides into either the base itself for near inspection/higher magnification, or into one or both of the “stackable” insert adaptors for greater working distance/lower magnification
Fits Mighty Scope 1.3M and 5M. Please note this cradle stand will not work with 500x model.
When using a base+single adaptor an entire USA penny can be viewed, and while using the base+both adaptors an entire USA quarter can be viewed. The clear acrylic cutaway base design allows easy accessibility of tools and light to the observed object.