Bath Oil, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
An extremely stable, medium viscosity silicone heating bath oil
Will not show discoloration after 40 hours at maximum temperature.
Ordering information: Available in two temperature ranges: Low Temp — maximum 180°C; or High Temp — maximum 230°C. Low temp oil color is CLEAR. High temp oil color is AMBER.
Customers Who Bought This Also Bought