SP Bel-Art Wash Bottles, Needle Spray Narrow Mouth, LDPE, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
For an extra-fine, needle-point spray, use these easy-to-squeeze low-density polyethylene wash bottles.
- LDPE construction
- Extra-fine needle-point spray
- Easy to use
Needle spray allows for fine, precise dispensation of washing solution.
Customers Who Bought This Also Bought