SP Bel-Art Gas Washing Bottle with Fritware® Dispersion Disc, Bel-Art Products, a part of SP
# de catalogue 16404-701
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Capacity:250 ml (8.5 oz.)
- Description:Gas Washing Bottle
- Height:27.3 cm (103/4")
- Cat. no.:16404-701
- Supplier No.:110300000
Spécifications
A propos de cet article
Graduated 250 ml polypropylene cylinder, with a stable extended base for use in precipitating elements from gas.
- Practical and ecnomical
- Coarse grade porosity disc
Gas is dispersed through a removable fitting that has a coarse grade porosity, polyethylene dispersion disc. The silicone stopper holds 6.3 mm (¹/₄") Ext.Ø tubes for entrance and exit of gas. Maximum pressure is 0.5 psi.
Customers Who Bought This Also Bought