SP Bel-Art Space-Saving Vented Jerrican, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
High-density polyethylene jerricans have a unique 20 cm (8") long spout with a built-in air vent for smooth pouring.
- Retractable spout
- Leakproof screw cap
- Extra hand grip provides positive control when pouring
- Graduated measurements
- Extra headspace for expansion of contents
The spout retracts into the threaded collar to conserve space, and the collar is sealed by a leakproof screw cap. An extra hand grip on the bottom gives positive control when pouring. The 5 gal. jerrican has a 2.5 cm (1") Int.Ø pouring spout; other sizes have 1.9 cm (³/₄") Int.Ø spouts. Graduated in U.S. gallons and liters, they feature extra headspace for expansion of contents.
Customers Who Bought This Also Bought