Upchurch Scientific® Gauge Plugs, IDEX Health & Science
Fournisseur: Upchurch Scientific
Certificats
A propos de cet article
These Upchurch Scientific® MicroTight™ Gauge Plugs are designed as accessories for MicroTight Unions and Adapters.
- MicroTight™ gauge plugs
- Designed as MicroTight accessories
- Assist in making zero-dead-volume connections
The plugs assist in making true zero-dead-volume connections by helping to ensure the tubing ends butt against each other inside the connection.
Customers Who Bought This Also Bought