Upchurch Scientific® Y Connectors, Multi-Port Connectors, IDEX Health & Science
Fournisseur: Upchurch Scientific
Certificats
A propos de cet article
Upchurch Scientific® PEEK™ Y Connectors are designed to split a stream or join two streams together, just like a tee
However, the configuration of a tee can lead to turbulent flow and solvent outgassing, which increases baseline noise and reduces sensitivity. The geometry of a Y connector creates much less turbulence and thus can improve analytical results. These standard Y Connectors use 1/4-28 Flangeless fittings.
Customers Who Bought This Also Bought