A propos de cet article
Upchurch Scientific® Quick Connect Luer Adapters by IDEX Health & Science adapt luers to 1/4-28, 10-32, 5/16-24, or M6 threaded ports.
- Designed to work in a variety of applications
- Each Quick Connect Luer Adapter conforms to ISO requirements for medical Luer taper configuration and performance
- Female Quick Connect Luer Adapters can be used with any of the Male Luers
Designed to work in a variety of applications, each Quick Connect Luer Adapter conforms to ISO requirements for medical Luer taper configuration and performance (45 psi / 3.1 bar)
Upchurch Scientific® LuerTight™ fittings by IDEX Health & Science are specifically designed to connect 1/16 in and 1/8 in OD tubing fluoropolymer tubing without barbs or nuts. By integrating ferrules into the Luer bodies, LuerTights reliably hold tubing in place while giving you the convenience of a Luer connection. LuerTight fittings provide a quick and easy disconnection option, and are less bulky and more economical than nut-to-Luer style fittings.
Note: The Female Quick Connect Luer Adapters can be used with any of the Male Luers, i.e., those with and without lock hubs.