Four Compartment Dispenser With Front Openings, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Dispenser with 4 compartments and front acces for gloves, bouffants, etc
Wall mounts-low profile.
Spécifications
- Catalog No:
- 98106-930
- 98106-896
- PPE type:
- Multi-use
- Multi-use
- Material:
- PETG
- Acrylic
- No. of compartments:
- 4
- 4
- Dispenser type:
- Wall-mountable
- Wall-mountable
- Thickness:
- 6 mm (0.25")
- 6 mm (0.25")