One Compartment Dispenser With Front Opening, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
A robust dispenser that can be wall mounted and is deep enough to be sturdy on a Metro rack or shelf.
- Front access hole
- Sloped lid with full length piano hinge
- Mounts to wall or free standing
- Flame polished edges for a smooth finish
Available in Acrylic or PETG (PETG is safe with IPA).
Spécifications
- Catalog No:
- 89140-744
- 89140-746
- PPE type:
- Multi-use
- Multi-use
- Material:
- Acrylic
- PETG
- No. of compartments:
- 1
- 1
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable