3 Compartment Dispenser, S-Curve Technologies
Supplier: S-Curve Technologies
Certificates
About this item
Mid-size dispenser with 3 compartments but low profile on wall.
Specifications
- Catalog No:
- 87000-014
- 89208-702
- PPE type:
- Multi-use
- Multi-use
- Material:
- Acrylic
- PETG
- No. of compartments:
- 3
- 3
- Dispenser type:
- Bench top/wall-mountable
- Bench top/wall-mountable
- Thickness:
- 6 mm (0.25")