Dispenser With Front Access Tray, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
1 compartment dispenser with front tray-ideal for ear plugs, safety glasses, etc, wall mounts or free standing.
Spécifications
- Catalog No:
- 87000-026
- 89208-680
- PPE type:
- Earplugs
- Earplugs
- Material:
- Acrylic
- PETG
- No. of compartments:
- 1
- 1
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Thickness:
- 6 mm (0.25")
- 6 mm (0.25")