A propos de cet article
Lucigen’s NxGen™ phi29 DNA Polymerase is designed for excellent strand displacement, fidelity, and speed
phi29 DNA Polymerase is a highly processive DNA polymerase with a powerful strand displacement activity and a 3′→ 5′ proofreading exonuclease function. The enzyme is capable of up to 70,000 insertions per binding event.
Performance Specifications: 1) Unit Concentration: 10,000 U/mL. 2) Purity (SDS-PAGE): >99%. 3) SS Exonuclease: Functional. 4) E. coli 16S rDNA Contamination: 100 U <10 copies. 5) Endonuclease: 100 U <10% converted. 6) Storage: -20°C.
Source: A recombinant E. coli strain carrying the phi29 DNA Polymerase gene from bacteriophage phi29.