Automation Pipette Tips for PatchXpress Workstations, Thermo Scientific
Catalog # 89025-942
Supplier: Thermo Scientific
Specifications
- Volume:50 µl
- Tip Color:Clear
- Packed:96 Tips/Tray, 10 Trays/case
- Cat. no.:89025-942
- Sterile:+
- For Use With:PatchXpress automated workstations
- Supplier No.:902-261XP
Specifications
About this item
Thermo Scientific provides a solution for your PatchXpress robotic pipette tips.
Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Customers Who Bought This Also Bought