Tornado Safe Rooms, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
SECURALL® Storm Shelters & Safe Rooms are built in accordance with specifications set forth by FEMA, the National Storm Shelter Association (NSSA), and the International Code Council (ICC).
- Multiple sizes available
- Manual close door types
- SMaRT Certified
Tornado shelters comply with the recommendations established in the documents: FEMA 320 - Taking Shelter from the Storm (written in conjunction with Texas Tech's Wind Engineering Research Center) and ICC-500. Securall Tornado Safe's FEMA 320/361/ICC-500 Proprietary Wall Assembly & FEMA 320/ICC-500 door assembly was tested and successfully passed missile impact resistance tests at Texas Tech University’s Wind Science and Engineering Research Center.
Certifications: SMaRT Certified; Built in Accordance with FEMA Standards.
Ordering information: Standard Color is Gray, please contact your Avantor Furniture Specialist for more information.
Spécifications
- Catalog No:
- 89237-664
- 89237-666
- 89237-668
- 89237-670
- 89237-672
- 89237-674
- 89237-676
- 89237-678
- 89237-680
- 89237-662
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified