Drum Spill Stations, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Controlling unwanted spills is easier than ever with SECURALL® Drum Spill Pallets
Built to provide a lifetime of service, these secondary containment Drum Spill Stations provide an easy and safe solution to organizing hazardous waste in drums. Rugged all-welded 12-gauge galvannealed steel offers long term use. Our Drum Spill Stations are specifically designed to be used as a holding area for drums, dispensing station or waste collection center. All units feature steel planking and forklift channels for easy mobility. Durable enough to be transported at full capacity.
Certifications: SMaRT Certified; Complies with EPA Spill Containment Requirements (40CFR264.175).
Ordering information: Standard Color is Gray, please contact your Avantor Furniture Specialist for more information.
Spécifications
- Catalog No:
- 89237-804
- 89237-806
- 89237-808
- 89237-810
- 89237-812
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Storage:
- 2 - Drums
- 4 - Drums
- 6 - Drums
- 9 - Drums
- 12 - Drums
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified