Stainless Steel Industrial Cabinets, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Made from 18 gauge steel with #4 brushed finish.
- Stainless steel 304 is corrosion and acid resistant to last a lifetime and handles everyday use in industrial environments
- Each shelf is easily adjustable and carries a 500 lbs. load capacity
- Flush-mounted stainless steel locking door handle eliminates projecting obstacles
Perfect for controlled environments and clean room applications such as hospitals and laboratories. Durable, 18-gauge stainless steel cabinets offer extra strength. Includes 3-point recessed stainless steel paddle handle with two brass keys and self-latching doors with steel rods.
Certifications: SMaRT certified.
Ordering information: Unpainted.
Spécifications
- Catalog No:
- 89237-816
- 89237-818
- 89237-820
- 89237-822
- 89237-824
- 89237-826
- 89237-828
- 89237-830
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified