Stannous chloride reagent II for phosphate analysis (with extraction)
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
:10025-69-1
Formule:C3H8O3
MW:92,09 g/mol
:Stannous chloride reagent II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Eclair:|Point eclaire:
:M.W. 92,09 g/mol
Point d'ébullition:~ 290 °C
Point de Fusion:18.2 °C
Densité:1.2637 g/cm³
Point d'éclair:199 °C
:Boiling Pt:
:Boiling Pt: ~ 290 °C
:Melting Pt:
:Melting Pt: 18.2 °C
:Flash Pt:
:Flash Pt: 199 °C
:M.W. 92.09 g/mol
:~ 290 °C
:199 °C
:GHS07
Température d'entreposage:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Résultats des tests de spécification
- Appearance:Clear, colorless, odorless
- Stannous chloride reagent II for phosphate analysis (with extraction):