ContactUsLinkComponent
Cyanide Standard, 1 ml = 1 mg CN, 1000 ppm CN, Ricca Chemical Company
Cyanide Standard, 1 ml = 1 mg CN, 1000 ppm CN, Ricca Chemical Company
Supplier:  Ricca Chemical
Certificates
Cyanide Standard, 1 ml = 1 mg CN, 1000 ppm CN, Ricca Chemical Company
Cyanide Standard, 1 ml = 1 mg CN, 1000 ppm CN, Ricca Chemical Company
Supplier:  Ricca Chemical
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.