Paint and Ink Storage Cabinets, SECURALL®
Paint storage cabinets for 1 gal. paint cans are available in 20, 40, 60 and up to 120 gal. capacities.
- Warning label reads "Flammable - Keep Fire Away"
- Perfect for print shops or auto body repair
Finished in two-part urethane paint.
Standard color is yellow, please contact your Avantor Furniture Specialist for more information.
Certifications: SMaRT Certified; FM Approved.
- Catalog No:
- 89237-730
- 89237-732
- 89237-734
- 89237-736
- 89237-738
- 89237-740
- 89237-742
- 89237-726
- 89237-728
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
Customers Who Bought This Also Bought