About this item
AcroPrep Advance 96-well filter plates are chemically compatible with harsh organic and / or aqueous solvents and contain inert materials of construction to provide a stable platform and ensure complete recovery of samples and low non-specific binding. The water wettable Polytetrafluoroethylene (wwPTFE) is a universally chemically compatible membrane and offers fast flow rates, low protein binding, and low levels of UV-absorbing extractables. These plates are suitable for applications such as metabolic studies, molecular or drug synthesis reactions, sample prep using solvents, and aggressive filtration applications.
- High performance wwPTFE membrane is a hydrophilic membrane with broad chemical compatibility, and excellent particle retention
- The PTFE (polytertrafluoroethylene) membrane is a hydrophobic membrane with broad chemical compatibility, and excellent particle retention
- AcroPrep Plates eliminate crosstalk between wells. Membranes are individually sealed in each well with a proprietary sealing process and each well has a fluid director
- AcroPrep Advance plates have a unique well geometry resulting in faster, more uniform filtration rates across the plate with reduced hold-up volume
- AcroPrep Advance plates have an optimized outlet tip providing direct flow of samples into receiver plate without concerns of cross-contamination
- AcroPrep Advance plates have a smooth top surface and textured window allow for easy labeling on the plates
AcroPrep Advance filter plates with Next Generation wwPTFE (hydrophillic polytetrafluoroethene) membranes are an all-purpose, hydrophilic membrane for aqueous, acidic, basic, non-aggressive organic, and aggressive organic solutions. The wwPTFE membrane offers low protein binding and low levels of UV-absorbing extractables. For applications requiring a hydrophobic membrane the PTFE (polytertrafluoroethylene) membrane has broad chemical compatibility, and excellent particle retention.
AcroPrep plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid. The robotic-friendly design has single-piece construction, designed in accordance with the ANSI/SLAS X-2004 standards. Plates are stackable with and without lids. A serialized barcode label allows the use of automated tracking systems and identifies the membrane type.
Recommended centrifugation speed: 1500×g (AcroPrep™ Advance 96-Well Plates); 1000 - 3000×g (AcroPrep™ 96-Well Plates)
Recommended operating vacuum: >25.4 cm Hg (10" Hg)
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA89137-184
- CA89137-186
- CA89135-692
- CA76423-870
- 76308-650
- 76308-652
- CA76423-868
- CA89135-694
- CA-76308-646
- CA76308-642
- CA76423-864
- CA89135-710
- 76308-644
- CA89135-712
- 76308-648
- CA-28148-662
- 89137-184
- 28148-664
- Typical hold-up volume:
- 14 μl: Vacuum; 2 μl: Centrifuge
- 15 μl: Vacuum; 1 μl: Centrifuge
- 14 μl: Vacuum; 2 μl: Centrifuge
- 15 μl: Vacuum; 1 μl: Centrifuge
- 14 μl: Vacuum; 2 μl: Centrifuge
- 15 μl: Vacuum; 1 μl: Centrifuge
- 14 μl: Vacuum; 2 μl: Centrifuge
- Well bottom area:
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²