Laboratory Extra Storage Shelves, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Unique storage shelf can add space in any lab.
- Flame polished edges for a smooth finish
- Wall mounts or free-standing
- ¹/₄" acrylic or PETG material
Three shelves for papers, labels, pipets, pens, gloves, etc.
Customers Who Bought This Also Bought