Beta Storage Container, S-Curve Technologies
# de catalogue 14007-132
Fournisseur: S-Curve Technologies
Spécifications
- Dimensions:304.8×250×250 mm (12×10×10")
- Description:Storage Container, 8 mm (3/8") acrylic, clear
- Cat. no.:14007-132
- Supplier No.:BC-502
Spécifications
A propos de cet article
Lab container for beta storage, table top only.
Customers Who Bought This Also Bought