These lockable gas storage cabinets limit access to authorized personnel, protecting cylinders from theft, damage and/or tampering.
Healthcare organizations such as hospitals, nursing homes, and emergency medical services will be prepared for their 3-year JCAHO accreditation survey when their oxygen cylinders are stored safely in SECURALL® MedGas Cabinets. SECURALL® proudly manufactures the highest quality safety storage products available. SECURALL® Gas Cylinder Storage Cabinets are available for storage of D, E, and H cylinders in varying capacities to meet any storage need. The "Capacity_NA" represents Storage capacity. The "Storage" represents Storage types.
The heavy-gauge, double-walled construction shields your cylinders from heat, sparks and flame, while separating them from flammable and combustible materials. Designed for D- and E-size cylinders up to 33-3/4" long x 4-1/4" diameter, and H-size cylinders up to 60” long x 9-1/4” diameter.
- Catalog No:
- 89237-686
- 89237-708
- 89237-688
- 89237-710
- 89237-690
- 89237-712
- 89237-692
- 89237-714
- 89237-694
- 89237-716
- 89237-696
- 89237-718
- 89237-698
- 89237-720
- 89237-700
- 89237-722
- 89237-702
- 89237-724
- 89237-682
- 89237-704
- 89237-684
- 89237-706
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified