ContactUsLinkComponent
LaMotte® AP Environmental Science Water Quality Assessment Module
LaMotte® AP Environmental Science Water Quality Assessment Module
# de catalogue 470136-872
Fournisseur:  LaMotte
LAMOTTE AP ENV SCI WATER QUALITY MODULE
LaMotte® AP Environmental Science Water Quality Assessment Module
# de catalogue 470136-872
Fournisseur:  LaMotte
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    LaMotte AP Environmental Science Water Quality Assessment Module
  • Cat. no.:
    470136-872
  • Supplier No.:
    5845
  • No. of Parts:
    10 tests
Spécifications
A propos de cet article
A Comprehensive Water Pollution Test Kit
This complete activity for your APES course is designed to satisfy water quality analysis requirements. It includes an extensive curriculum that uses the exploration of the Water Quality Index to teach students STEM-based skills that they will apply through classroom and field activities to satisfy Section VI (Water Pollution) of the AP Environmental Topics Outline.
Kit Contents
: Ten nutrient - TTC/MacConkey BioPaddles for coliform testing; Salt/TDS/Temp Tracer for temperature and TDS determination; Teacher Resource CD-ROM containing Teacher and Student Guides, water quality test procedures, PowerPoint presentations, and Quick Time iPad/iPod videos for STEM design activities and STEM skills, Watershed Ecology, WQI, Point/Non-Point Pollution, glossary and free app; Free LaMotte BioPaddle colony; Identification app module for use with the Water Quality Educator.
  • Curriculum to teach assessment of water quality
  • Aligned to NGSS HS-LS2-1, HS-LS2-7, HS-ESS2-2, HS-ESS2-5, HS-ESS3-1, HS- ESS3-6
  • Operating Temperature 35 °C
This complete activity for your APES course is designed to satisfy water quality analysis requirements. It includes an extensive curriculum that uses the exploration of the Water Quality Index to teach students STEM-based skills that they will apply through classroom and field activities to satisfy Section VI (Water Pollution) of the AP Environmental Topics Outline.
Kit Contents: Ten nutrient - TTC/MacConkey BioPaddles for coliform testing; Salt/TDS/Temp Tracer for temperature and TDS determination; Teacher Resource CD-ROM containing Teacher and Student Guides, water quality test procedures, PowerPoint presentations, and Quick Time iPad/iPod videos for STEM design activities and STEM skills, Watershed Ecology, WQI, Point/Non-Point Pollution, glossary and free app; Free LaMotte BioPaddle colony; Identification app module for use with the Water Quality Educator.
Customers Who Bought This Also Bought