Earth's Interior Globe
# de catalogue 470136-882
Fournisseur: VWR International
Spécifications
- Description:Earth's Interior Globe
- Cat. no.:470136-882
- Supplier No.:BD0078
Spécifications
A propos de cet article
Get an Inside Look at Our World
Every student will be in awe of our planet with this beautifully hand-painted resin model in your classroom. It depicts the internal structure of the earth, featuring seven detachable parts that illustrate the crust, mantle, lithosphere, mesosphere, asthenosphere, and inner and outer core. Size: 9" diameter globe, mounted on a sturdy metal tripod base.
Every student will be in awe of our planet with this beautifully hand-painted resin model in your classroom. It depicts the internal structure of the earth, featuring seven detachable parts that illustrate the crust, mantle, lithosphere, mesosphere, asthenosphere, and inner and outer core. Size: 9" diameter globe, mounted on a sturdy metal tripod base.
Customers Who Bought This Also Bought