Bottle Holder, S-Curve Technologies
# de catalogue 89140-732
Fournisseur: S-Curve Technologies
Spécifications
- Description:2-bottle holder PETG (6 mm/1/4"), mounts to wall
- Cat. no.:89140-732
- L×W×H:22.8×13.1×10.2 cm (9×5×4")
- Supplier No.:BH-200 PETG
Spécifications
A propos de cet article
Holder for 2 bottles, wall mounts, screws included
Customers Who Bought This Also Bought