Wiper Dispenser for Presaturated Wipes, S-Curve Technologies
# de catalogue 89140-774
Fournisseur: S-Curve Technologies
Spécifications
- Material:PETG, 6 mm (¼"), safe with IPA
- Dimensions:240.5W x 101.6H x 161.2D mm (9.5 x 4 x 6.5")
- Supplier No.:WDL-1048PETG
Spécifications
A propos de cet article
Table top dispenser for presaturated wipes
Customers Who Bought This Also Bought