Three Compartment Multi-Purpose Dispenser, S-Curve Technologies
Supplier: S-Curve Technologies
Certificates
About this item
Dispenser with 3 compartments for gloves, bouffants, etc.
Wall mount or free-standing.
Specifications
- Catalog No:
- 98106-928
- 87000-012
- PPE type:
- Multi-use
- Multi-use
- Material:
- PETG
- Acrylic
- No. of compartments:
- 3
- 3
- Dispenser type:
- Wall-mountable
- Wall-mountable
- Thickness:
- 6 mm (0.25")
- 6 mm (0.25")