Bouffant and Beard Cover Dispenser, S-Curve Technologies
Catalog # 89208-674
Supplier: S-Curve Technologies
Specifications
- Dispenser type:Bench top
- Material:PETG
- No. of compartments:1
- PPE type:Multi-use
- Color:Clear
- Description:Dispenser
- Thickness:3 mm
- Cat. no.:89208-674
- Supplier No.:BD-700 PETG
Specifications
About this item
Dispenser with rectangular opening for product access.
Customers Who Bought This Also Bought