Front Access Storage Bin Dispenser, S-Curve Technologies
# de catalogue 89208-708
Fournisseur: S-Curve Technologies
Spécifications
- Color:Clear
- Description:Dispenser with one compartment and front access
- Cat. no.:89208-708
- W×D×H:42×30.5×43 cm (16.5×12×17")
- Supplier No.:SBD-19 PETG
Spécifications
A propos de cet article
One compartment dispenser made from PETG.
- PETG is safe with IPA
- Flame treated edges for a smooth finish
- Keyholes on back for wall mounting
Designed for large volumes of gloves, shoe covers, bouffants, etc.
Customers Who Bought This Also Bought