Safety Glass Dispenser, 15 compartments, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
This safety glass dispenser holds up to 15 pairs of glasses.
- Mounts to wall vertically or horizontally or free standing
- Keeps glasses clean and safe
- Flame treated edges for smoothness
Made from PETG or acrylic.
Spécifications
- Catalog No:
- 32948-861
- 89208-716
- PPE type:
- Safety glasses
- Safety glasses
- Material:
- Acrylic
- PETG
- No. of compartments:
- 15
- 15
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Thickness:
- 6 mm (0.25")
- 6 mm (0.25")