Spécifications
- Description:Hamilton Model 1001 LT Pipette Controller Syringe
- Cat. no.:89221-468
- Needle Length:No Needle Included
- Point Style:No Needle Included
- Gauge:No Needle Included
- Supplier No.:84001
- Termination:LT
Spécifications
A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes and accessories in the industry
Pipette Controller Syringes are intended to be used in place of rubber bulbs to easily and precisely fill glass pipettes. The free-sliding plunger coarsely fills the pipette, and the knurled plunger screw completes fine volume adjustment. The Luer tip syringe termination allows easy connection of tubing to glass pipettes.
Terminations
Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
Each is manufactured to achieve the highest level of accuracy and precision possible.