Beta Tool™, Tubing Straightener, MicroSolv
Catalogue # 97051-378
Fournisseur: MicroSolv Technology Corp
Spécifications
- Description:Tubing straightener
- Material:Stainless steel
- Cat. no.:97051-378
- Supplier No.:46010-BT
Spécifications
À propos de cet article
Beta Tool™ is a pair of precision, high grade pliers and can be used as needle nose pliers or as a tubing straightener.
- Use to remove kinks or bends in standard outer diameter HPLC stainless steel tubing
It brings 1/16" outer dia. stainless steel tubing back to a perfect round without compromising the inner diameter.