Magnetic Spin Vanes for NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These fluoropolymer resin spin vanes have cross-mounted molded-in magnetic bars for V-vials.
These exceptionally sturdy vials have a conical interior bottom that provides total downward drainage for maximum retrieval of contents by syringe or micropipette.
Customers Who Bought This Also Bought