About this item
Rechargeable -26P Series UV Lamps offer versatility and variety in a powerful 6/12 watt rechargeable design. UVS-26P has two shortwave tubes. This model allows you to use two tubes at a time, or just one. UVL-26P has two longwave ultraviolet tubes. This model allows use of one or two tubes at a time. UVSL-26P has one shortwave and one longwave UV tube. This model allows use of one tube at a time. The toggle switch allows users to easily select longwave or shortwave UV.
Lamps are durable and can be used for both field and indoor/lab use
Each lamp is powered by a built-in lead acid battery which is charged from a standard wall outlet. The wall cord is for charging only and not for powering the lamp.
An optional 12V adapter operates the lamp from a car cigarette lighter.
The C-10P Cabinet can be used with the Portable and Portable/Rechargeable UV Lamps for fluorescing materials in a darkened area.