Boerner MicroTest Slide. Cs12, Wheaton
# de catalogue 48351-005
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Description:Boerner micro test slide
- Cat. no.:48351-005
- Supplier No.:798550
Spécifications
A propos de cet article
The WHEATON Boerner Micro Test Slide is ideal for the Boerner-Jones-Lukens microflocculation test
The slide is used for precedures involving 0.01 to 0.15mL of fluid. The slide has 10 cells arranged in 2 rows of 5 each. Each cell is 2.5mm deapp and is designed to produce flattned drops of uniform depth. The slide is manufactured from soda-lime glass.
Customers Who Bought This Also Bought