ContactUsLinkComponent
Alcohol Burner, WHEATON, DWK Life Sciences
Alcohol Burner, WHEATON, DWK Life Sciences
# de catalogue 16299-098
Small Business Enterprise
Alcohol BurnerWith Ground Glass Cap, 100mL
Alcohol Burner, WHEATON, DWK Life Sciences
# de catalogue 16299-098
Small Business Enterprise
Spécifications
  • Volume:
    100 ml (3.5 oz.)
  • Description:
    Alcohol burner, 100 ml, Pkg of 6
  • Supplier No.:
    237070
Spécifications
A propos de cet article
The WHEATON Alcohol Burner is manufactured from soda-lime glass and is designed for use with isopropyl or denatured ethyl alcohol
  • The burner's low heat flame is well suited for microscopy purposes
  • Reservoir holds 100 mL of alcohol
  • Glass cap is used to reduce evaporation of alcohol when not in use
Dia.× H: 75×100 mm
Customers Who Bought This Also Bought