Veterinary Needles, Cadence Science®
Fournisseur: Avantor
Certificats
A propos de cet article
These hypodermic needles are all fitted with a large knurled nickel plated hub which allows for secure gripping even under the most difficult circumstances.
Ordering information: Non-sterile. Packaged 12 needles per box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.