O-Rings, EPDM, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Ethylene-Propylene (EPDM) o-rings
EPDM is a type of synthetic rubber of ethylene-propylene, and is also referred to as EPDM; E = ethylene; P = propylene; D = diene; and M refers to monomer, or M-class rubber (ASTM D1418). EPDM features a good, soft general seal with wide applications, and good compression set. Can be green, brown, or black in color. USP Class VI material.
Customers Who Bought This Also Bought